Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0229

Sigma-Aldrich

PF-03246799 hydrochloride

≥98% (HPLC)

Synonym(s):

6,7,8,9-Tetrahydro-2-(phenylmethyl)-5H-pyrimido[4,5-d]azepine hydrochloride, PF-3246799 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N3 · xHCl
CAS Number:
Molecular Weight:
239.32 (free base basis)
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

C1(CC2=NC(CCNCC3)=C3C=N2)=CC=CC=C1.Cl

InChI

1S/C15H17N3.ClH/c1-2-4-12(5-3-1)10-15-17-11-13-6-8-16-9-7-14(13)18-15;/h1-5,11,16H,6-10H2;1H

InChI key

ICVPRIZGSIUCOE-UHFFFAOYSA-N

Biochem/physiol Actions

PF-3246799 is a potent serotonin 5-HT2C receptor agonist with an EC50 of 4.5 nM and minimal activity at 5-HT2A and 5-HT2B receptors. PF-3246799 had good efficacy in a preclinical canine model of stress urinary incontinence.
PF-3246799 is a potent serotonin 5-HT2C receptor agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service