PIC0001
ReadyShield® Protease Inhibitor Cocktail
for use with fungal and yeast extracts, Non-freezing solution
Synonym(s):
Protease inhibitor cocktail
About This Item
Recommended Products
description
Fungal and Yeast Extracts
form
(non-freezing liquid formulation )
quality
200
packaging
pkg of 1 mL (in amber polypropylene vial)
pkg of 5 mL (in amber polypropylene vial)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Preparation Note
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service