PB10100707CN
Pop-Bio ′Vü-F/C Imaging System Combo (both systems)
Synonym(s):
Imaging system, Pop-Bio ′Vü-F/C Imaging System Combo (both systems)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122600
Recommended Products
AC/DC input
24 V
packaging
pkg of 1 ea
Looking for similar products? Visit Product Comparison Guide
Application
Western blot and fluorescence gels
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service