MTCT9019
MTC™ Bio Standard Cap for FlowTubes™
non-sterile
Synonym(s):
culture cap, tube caps
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122500
NACRES:
NB.11
Recommended Products
sterility
non-sterile
packaging
bag of 8 × 125 ea
manufacturer/tradename
MTC™ Bio T9019
General description
Standard Cap for FlowTubes™, non-sterile, 125/bag, 1000/cs
Application
- Fluorescence activated cell sorting
- Flow cytometry
Features and Benefits
- 2 position culture caps
- For use with 12 x 75mm Flowtubes
Legal Information
FlowTubes is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service