Key Documents
MTCP1001PCR
MTC Bio Pure.Amp Transparent Sealing Film
to fit standard PCR plates
Synonym(s):
plastic tube, plate sealing film, sealing membrane
About This Item
UNSPSC Code:
41122410
NACRES:
NB.15
packaging
bag of 10 × 10 ea
manufacturer/tradename
MTC Bio P1001-PCR
General description
Sealing film, PCR 96 Well Plate Sealing Membrane, 10 bags of 10 seals, 100/pk
Features and Benefits
  • RNase, DNase, DNA, and PCR inhibitor free
  • Evaporation rates ≤ 5%
  • Inner packs of 10 membranes
  • -40° to 120°C
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)