MTCM5431
MTC™ Bio T-Shape Spreader
L × W 38 mm × 140 mm, sterile, pack of 500 ea (50 x bags 10 ea)
Synonym(s):
cell spreader
All Photos(1)
About This Item
UNSPSC Code:
41122103
NACRES:
NB.17
Recommended Products
sterility
sterile
packaging
pack of 500 ea (50 x bags 10 ea)
manufacturer/tradename
MTC™ Bio M5431
L × W
38 mm × 140 mm
General description
Spreader, T-Shape, 34 x 140mm, 50 sterile peel bags of 10 spreaders, 500/pk
Features and Benefits
- Spreads cultures and liquids evenly
- Ideal for bacterial CFU counts
- Smooth edges - minimizes gouging
- Pre-sterilized - no flaming required
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)