Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK492

Sigma-Aldrich

Fumarate Assay Kit

new

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

application(s)

food and beverages

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

General description

Fumarate, or fumaric acid, is one of the key components in the TCA cycle and is used by cells to form ATP. Human skin, when exposed to sunlight, will naturally produce fumaric acid. Fumarate is used as an additive by the food and beverage industry. Fumaric acid esters are also used to treat psoriasis. Increased urinary fumarate may be due to impaired Krebs Cycle function, a defect in the enzyme fumarase, or mitochondrial function.

Features and Benefits

  • Linear detection range 0.005 to 2 mM fumarate.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the determination of fumarate in food, beverage, and other biological samples (e.g. cell lysate, tissue homogenate, and serum).

Principle

The Fumarate Assay Kit is based on the fumarase catalyzed hydration of fumarate to malate. The malate is then oxidized by malate dehydrogenase generating NADH, which reduces a formazan (MTT) dye. The intensity of the product color, measured at 565 nm, is proportional to the fumarate concentration in the sample.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service