Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA023924

Sigma-Aldrich

Anti-RPAIN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-MGC4189, Anti-RIP, Anti-RPA interacting protein, Anti-hRIP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

YNLRITSGVVVCQCGLSIPSHSSELTEQKLRACLEGSINEHSAHCPHTPEFSVTGGTEEKSSLLMSCLACDTWAVI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPAIN(84268)

General description

RPAIN (replication protein A interacting protein), also known as RIP (RPA interacting protein), is an adaptor protein, that is transcribed into 9 isoforms as obtained from a premade human fetal brain library. The longest isoform is hRIPa, which is composed of 219 amino acids. It contains an arginine-rich basic region in its N-terminal, succeeded by an acidic region and two Zn finger-like domains in its C-terminal. hRIPβ lacks one Zn finger-like region. RPAIN gene is localized to human chromosome 17p13 and is composed of seven exons.

Immunogen

RPA interacting protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RPAIN (replication protein A interacting protein) functions as a linker between RPA (replication protein A) protein and importin β. This complex between the a isoform of RPAIN and importin β facilitates the transport of RPA into the nucleus. In cervical cancer cells, the a form is elevated, where it controls cell proliferation through the regulation of RPA transport. RPAINβ resides in the promyelocytic leukemia (PML) nuclear bodies, where it is responsible for the transportation of RPA into the PML bodies. It is also responsible for the release of RPA upon UV (ultraviolet) irradiation. This protein promotes the IRES (internal ribosome entry site)-mediated translation of HIV-1 (human immunodeficiency virus).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75599

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junsoo Park et al.
Molecular and cellular biology, 25(18), 8202-8214 (2005-09-02)
Replication protein A (RPA) is a single-stranded-DNA-binding protein composed of three subunits with molecular masses of 70, 32, and 14 kDa. The protein is involved in multiple processes of eukaryotic DNA metabolism, including DNA replication, repair, and recombination. In Xenopus
Jin-Zhong Chen et al.
DNA and cell biology, 24(7), 464-469 (2005-07-13)
XRIP alpha was identified as an adapter protein involved in RAP nuclear import. Several homologs were reported in mammal EST analysis, but the expression pattern and genomic organization of hRIP isoforms were not clarified yet. We isolated nine isoforms of
Jinfeng Liu et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 6(2), 308-321 (2011-03-02)
Nuclear export and translation of HIV-1 RNA are two important posttranscriptional events for HIV-1 gene expression and replication. HIV-1 Rev functions to export unspliced and incompletely spliced HIV-1 RNA from the nucleus to the cytoplasm; it requires interaction with several
Sim Namkoong et al.
FEBS letters, 586(20), 3753-3760 (2012-09-27)
Replication protein A (RPA) is a eukaryotic single-stranded DNA binding protein that is essential for DNA replication, repair, and recombination, and human RPA interacting protein α (hRIPα) is the nuclear transporter of RPA. Here, we report the regulatory role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service