Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

HPA021995

Sigma-Aldrich

Anti-ECHS1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Enoyl-CoA hydratase 1, Anti-Enoyl-CoA hydratase, mitochondrial, Anti-SCEH, Anti-Short chain enoyl-CoA hydratase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

ANFEYIIAEKRGKNNTVGLIQLNRPKALNALCDGLIDELNQALKIFEEDPAVGAIVLTGGDKAFAAGADIKEMQNLSFQDCYSSKFLKHWDHLTQVKKPVIAAVNGYAFGGGCELAMM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ECHS1(1892)

General description

ECHS1 (Enoyl CoA hydratase, short chain, 1, mitochondrial) is structurally located in chromosome 10q26.2-q26.3 region consisting of eight exons, with exons I and VIII with the 5′- and 3′-untranslated regions. It has a GC-rich 5′-flanking region with multiple copies of the SP1 binding motive. Its expressions have been found in the human liver, fibroblasts and muscles.

Immunogen

Enoyl-CoA hydratase, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ECHS1 (Enoyl CoA hydratase, short chain, 1, mitochondrial) is involved in the mitochondrial β-oxidation of fatty acid by catalyzing the second step of the process i.e. hydration of 2-trans-enoyl-coenzyme A (CoA) to L-3-hydroxyacyl-CoAs. It also participates in different metabolic pathways involving fatty acids and amino acids, valine. Study reports that heterozygous mutation in ECHS1 causes Leigh syndrome with hypotonia, metabolic acidosis, and developmental delay symptoms.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75487

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymes of fatty acid metabolism. II. Properties of crystalline crotonase.
J R STERN et al.
The Journal of biological chemistry, 218(2), 985-1002 (1956-02-01)
U Janssen et al.
Genomics, 40(3), 470-475 (1997-03-15)
The second step in mitochondrial fatty acid beta-oxidation is catalyzed by short chain enoyl-CoA hydratase (ECHS1; EC 4.2.1.17). Inherited disorders of this pathway of energy metabolism present clinical and laboratory features resembling sudden infant death syndrome and Reye-like syndrome. To
M Kanazawa et al.
Enzyme & protein, 47(1), 9-13 (1993-01-01)
Short chain enoyl-CoA hydratase (SCEH) catalyzes the second step of the mitochondrial fatty acid beta-oxidation spiral. We isolated cDNA clones for human SCEH to facilitate investigation of the enzyme structure of the gene and to examine the genetic background of
Chika Sakai et al.
Human mutation, 36(2), 232-239 (2014-11-14)
The human ECHS1 gene encodes the short-chain enoyl coenzyme A hydratase, the enzyme that catalyzes the second step of β-oxidation of fatty acids in the mitochondrial matrix. We report on a boy with ECHS1 deficiency who was diagnosed with Leigh

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service