Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA019684

Sigma-Aldrich

Anti-NOV antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Nephroblastoma overexpressed gene protein homolog, Anti-NovH, Anti-Protein NOV homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SCCLVCARQRGESCSDLEPCDESSGLYCDRSADPSNQTGICTAVEGDNCVFDGVIYRSGEKFQPSCKFQCTCRDGQIGC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOV(4856)

General description

NOV (Nephroblastoma overexpressed) is an extracellular matrix-associated protein belonging to the CCN (Connective tissue growth factor (CTGF), Cystein rich protein (Cyr61), and Nephroblastoma overexpressed gene (nov)) family. It consists of an N-terminal secretory signal and four domains with different functional attributes. It is expressed in kidney, heart, liver, and spleen.

Immunogen

Protein NOV homolog Precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73994

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Gellhaus et al.
The Journal of biological chemistry, 279(35), 36931-36942 (2004-06-08)
The gap junction protein connexin43 (Cx43) is thought to be involved in growth control in several tissues. Using the doxycycline inducible tet-on system, we generated human malignant trophoblast Jeg3 cells transfected with either Cx40, Cx43, or C-terminal truncated Cx43 (trCx43).
Cristiane G Lin et al.
The Journal of biological chemistry, 280(9), 8229-8237 (2004-12-22)
The novel angiogenic inducer CCN3 (NOV, nephroblastoma overexpressed) is a matricellular protein of the CCN family, which also includes CCN1 (CYR61), CCN2 (CTGF), CCN4 (WISP-1), CCN5 (WISP-2), and CCN6 (WISP-3). CCN3 is broadly expressed in derivatives of all three germ
Li-Ching Fan et al.
PloS one, 11(9), e0162933-e0162933 (2016-09-15)
In addition to contraction, myoepithelia have diverse paracrine effects, including a tumor suppression effect. However, certain myoepithelial markers have been shown to contribute to tumor progression. Transforming growth factor-β (TGF-β) is involved in the transdifferentiation of fibroblasts to contractile myofibroblasts.
Bruce L Riser et al.
The American journal of pathology, 184(11), 2908-2921 (2014-09-07)
Fibrosis is at the core of the high morbidity and mortality rates associated with the complications of diabetes and obesity, including diabetic nephropathy (DN), without any US Food and Drug Administration-approved drugs with this specific target. We recently provided the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service