Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA017919

Sigma-Aldrich

Anti-AKR1A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alcohol dehydrogenase [NADP+], Anti-Aldehyde reductase, Anti-Aldo- keto reductase family 1 member A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KYALSVGYRHIDCAAIYGNEPEIGEALKEDVGPGKAVPREELFVTSKLWNTKHHPEDVEPALRKTLADLQLEYLDLYLMH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKR1A1(10327)

General description

Aldo-keto reductase family 1 member A1 (AKR1A1) belongs to the aldo-keto reductase superfamily, and is expressed in the liver and kidney. The gene encoding it is localized on human chromosome 1p32-p33.

Immunogen

Alcohol dehydrogenase [NADP+] recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Aldo-keto reductase family 1 member A1 (AKR1A1) catalyzes the reduction of aliphatic aldehydes, aromatic aldehydes and biogenic amines. These reactions need nicotinamide adenine dinucleotide phosphate (NADPH). Studies have shown that AKR1A1 reduces 4-nitrobenzaldehyde and succinic semialdehyde in vitro. Mutation in the gene encoding this enzyme has been associated with heightened risk of non-Hodgkin′s lymphoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74597

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Fujii et al.
Cytogenetics and cell genetics, 84(3-4), 230-232 (1999-07-07)
Genomic DNA encoding for human aldehyde reductase (AKR1A1), a member of the aldo-keto reductase superfamily, was isolated and characterized. The genomic DNA is approximately 16 kb in length and contains eight exons which encode the entire coding region and the
Onkar S Bains et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(5), 904-910 (2008-02-16)
Aldo-keto reductases (AKRs) are a class of NADPH-dependent oxidoreductases that have been linked to metabolism of the anthracyclines doxorubicin (DOX) and daunorubicin (DAUN). Although widely used, cardiotoxicity continues to be a serious side effect that may be linked to metabolites
T O'connor et al.
The Biochemical journal, 343 Pt 2, 487-504 (1999-10-08)
Complementary DNA clones encoding human aflatoxin B(1) aldehyde reductase (AKR7A2), aldehyde reductase (AKR1A1), aldose reductase (AKR1B1), dihydrodiol dehydrogenase 1 (AKR1C1) and chlordecone reductase (AKR1C4) have been expressed in Escherichia coli. These members of the aldo-keto reductase (AKR) superfamily have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service