Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA016702

Sigma-Aldrich

Anti-KCNJ15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATP-sensitive inward rectifier potassium channel 15, Anti-Inward rectifier K(+) channel Kir4.2, Anti-Kir13, Anti-Potassium channel, inwardly rectifying subfamily J member 15

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SAVCQSRTSYIPEEIYWGFEFVPVVSLSKNGKYVADFSQFEQIRKSPDCTFYCADSEKQQLEEKYRQEDQRERELRTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ15(3772)

Immunogen

ATP-sensitive inward rectifier potassium channel 15 recombinant protein epitope signature tag (PrEST)

Application

Anti-KCNJ15 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72924

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Liu et al.
Cancer management and research, 11, 1211-1220 (2019-02-26)
KCNJ15 belongs to the inwardly rectifying potassium channel (KIR) family. Although members of the KIR family have been proven to play important roles in a variety of developmental processes, the molecular role and clinical effects of KCNJ15 in cancers remain
Marie-Christin Beer et al.
Biomolecules, 12(6) (2022-06-25)
Dedifferentiation and proliferation of retinal pigment epithelial (RPE) cells are characteristics of retinal diseases. Dedifferentiation is likely associated with changes of inwardly rectifying potassium (Kir) channels. The roles of Kir4.2 channels in viability, and proliferation of cultured RPE cells were
Akikazu Fujita et al.
The Journal of physiology, 540(Pt 1), 85-92 (2002-04-03)
Hydrochloric acid (HCl) is produced in parietal cells of gastric epithelium by a H(+)-K(+) pump. Protons are secreted into the gastric lumen in exchange for K(+) by the action of the H(+)-K(+)-ATPase. Luminal K(+) is essential for the operation of
P Gosset et al.
Genomics, 44(2), 237-241 (1997-09-23)
The Down syndrome chromosome region-1 (DCR1) on subband q22.2 of chromosome 21 is thought to contain genes contributing to many features of the trisomy 21 phenotype, including dysmorphic features, hypotonia, and psychomotor delay. Isolation, mapping, and sequencing of trapped exons
Wenjun He et al.
American journal of physiology. Gastrointestinal and liver physiology, 301(4), G591-G600 (2011-07-02)
Potassium ions are required for gastric acid secretion. Several potassium channels have been implicated in providing K(+) at the apical membrane of parietal cells. In examining the mRNA expression levels between gastric mucosa and liver tissue, KCNJ15 stood out as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service