Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA011762

Sigma-Aldrich

Anti-GALNT6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-GalNAc-T6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LIMYSCHGLGGNQYFEYTTQRDLRHNIAKQLCLHVSKGALGLGSCHFTGKNSQVPKDEEWELAQDQLIRNSGSGTCLTSQDKKPAMAPCNPSDPHQLWLFV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALNT6(11226)

Immunogen

polypeptide N-acetylgalactosaminyltransferase 6

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GALNT6 (UDP-N-acetyl-α-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6) is a glycosyltransferase essential for the protection and maintenance of a healthy ocular surface. It is specifically expressed in the conjunctival goblet cells. GALNT6 is majorly involved in the initial step of O-glycosylation. During O-glycosylation, it catalyzes the transfer of N-acetyl galactosamine (GalNAc) to serine and threonine residues through some enzymatic activity. It is associated with the aberrant glycosylation in gastric mucosa, intestinal metaplasia, and gastric carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72000

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaru Noda et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(18), 4468-4481 (2018-05-31)
Purpose: We aimed to discover glycosyltransferase gene (glycogene)-derived molecular subtypes of colorectal cancer associated with patient outcomes.Experimental Design: Transcriptomic and epigenomic datasets of nontumor, precancerous, cancerous tissues, and cell lines with somatic mutations, mismatch repair status, clinicopathologic and survival information
Pablo Argüeso et al.
Investigative ophthalmology & visual science, 44(1), 86-92 (2002-12-31)
It has been hypothesized that the biosynthesis of O-linked glycans on proteins, particularly on the highly O-glycosylated mucins, by the corneal and conjunctival epithelium is necessary for the protection and maintenance of a healthy ocular surface. The initial step in
Joana Gomes et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 57(1), 79-86 (2008-10-16)
Aberrant mucin O-glycosylation is often observed in cancer and is characterized by the expression of immature simple mucin-type carbohydrate antigens. UDP-N-acetyl-d-galactosamine:polypeptide N-acetylgalactosaminyltransferase-6 (ppGalNAc-T6) is one of the enzymes responsible for the initial step in O-glycosylation. This study evaluated the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service