Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA006903

Sigma-Aldrich

Anti-SPINT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HAI-1 antibody produced in rabbit, Anti-Hepatocyte growth factor activator inhibitor type 1 antibody produced in rabbit, Anti-Kunitz-type protease inhibitor 1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPINT1(6692)

Immunogen

Kunitz-type protease inhibitor 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SPINT1 (Serine peptidase inhibitor, Kunitz type 1) is a Kunitz-type serine protease inhibitor belonging to the Kunitz family. It is localized in the villous of cytotrophoblast (CTB). It controls the hepatocyte growth factor (HGF) activity by inhibiting HGF activator (HGFA). SPINT1 restricts the cancer metastasis by inhibiting the HGF activity. The interaction between SPINT1 and CTB may contribute in the villous differentiation and maintenance of the villous tree. It has been reported that SPINT1 also retards the activity of various pro-metastatic serine proteases associated with tumor spreading.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70169

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Mori et al.
Biology of reproduction, 76(1), 164-172 (2006-10-13)
Chorionic villi in the human placenta serve as essential structures in fetomaternal exchanges. According to the embryology and placentology literature, during the first trimester, the cytotrophoblast (CTB) layer that is subjacent to the syncytiotrophoblast (STB) and supported by a basal
Christian Parr et al.
International journal of cancer, 119(5), 1176-1183 (2006-03-25)
Hepatocyte growth factor (HGF) plays a plethora of roles in cancer metastasis and tumour growth. The interaction between tumour cells and their surrounding stromal environment is a crucial factor regulating tumour invasion and metastasis. Stromal fibroblasts are the main source
Andrei P Drabovich et al.
Molecular & cellular proteomics : MCP, 15(6), 2093-2107 (2016-04-14)
Estrogen receptor alpha (ERα)-mediated proliferation of breast cancer cells is facilitated through expression of multiple primary target genes, products of which induce a secondary response to stimulation. To differentiate between the primary and secondary target proteins of ERα signaling, we
T A Tervonen et al.
Oncogene, 35(14), 1832-1846 (2015-07-15)
Hepsin belongs to a family of cell-surface serine proteases, which have sparked interest as therapeutic targets because of the accessibility of extracellular protease domain for inhibitors. Hepsin is frequently amplified and/or overexpressed in epithelial cancers, but it is not clear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service