GE17-0855-01
Nick-column 20 ST
Cytiva 17-0855-01, pkg of 20 columns
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105501
NACRES:
NA.52
Recommended Products
packaging
pkg of 20 columns
manufacturer/tradename
Cytiva 17-0855-01
bed size
9 mm × 20 mm
column I.D.
9 mm
particle size
50-150 μm (dry)
capacity
100 μg binding capacity
Related Categories
General description
NICK Columns were originally designed for purification of nick-translated DNA fragments but are also suitable for separation of any labeled probe from unincorporated labeled nucleotides.
Application
NICK Columns were originally designed for purification of nick-translated DNA fragments but are also suitable for separation of any labeled probe from unincorporated labeled nucleotides. They are ready for immediate use, and operate by gravity flow. Tests show that at least 97% of radioactivity applied in a nick-translated mixture elutes in two well-separated peaks, corresponding to labeled DNA and unincorporated nucleotides, respectively. At least 90% of the applied DNA is recovered. Maximum sample volume is 100 μL.
Features and Benefits
- For rapid purification of labeled DNA (20 bases in length) from unincorporated radiolabeled nucleotides using gravity-fLow chromatography.
- Prepacked with Sephadex® G-50 DNA Grade (see illustra™ Sephadex® G-50 and G-100 DNA Grade) in distilled water with 0.15% Kathon CG/ICP Biocide.
- Can accommodate sample volumes up to 100 μL.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sephadex is a registered trademark of Cytiva
illustra is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service