EPM1335-0600
12 Port Gassing Manifold
for Eppendorf Brunswick Innova 42/42R
All Photos(1)
About This Item
UNSPSC Code:
41104405
NACRES:
NB.73
Recommended Products
manufacturer/tradename
Eppendorf® M1335-0600 (Global)
Related Categories
General description
The manifold delivers gas into the chamber and ultimately into the flasks themselves via twelve ports. You may elect to use splitters after the manifold to increase the number of ports for your application. Gassing manifolds are a factory fitted option and need specialist installation.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)