Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5495509100

Eppendorf® Rotor lid for rotor FA-18x2-KIT

Synonym(s):

Aerosol-Tight Rotor Lid for Centrifuge 5425/R, Aluminum Rotor Lid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5495509100

General description

Lid FA-18x2-KIT (Centr 5425/R)

Application

CENTRIFUGATION

Features and Benefits

aerosol-tight, aluminumfor Centrifuge 5425/5425R

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service