Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU158871

Sigma-Aldrich

MISSION® esiRNA

targeting human TRADD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAGGAGAACGAGCTCACCAGCCTGGCAGAGGACTTGCTGGGCCTGACCGATCCCAATGGCGGCCTGGCCTAGACCAGGGGTGCAGCCAGCTTTTGGAGAACCTGGATGGCCTTAGGGTTCCTTCTGCGGCTATTGCTGAACCCCTGTCCATCCACGGGACCCTGAAACTCCACTTGGCCTATCTGCTGGACCTGCTGGGGCAGAGTTGATTGCCTTCCCCAGGAGCCAGACCACTGGGGGTGCATCATTGGGGATTCTGCCTCAGGTACTTTGATAGAGTGTGGGGTGGGGGGGACCTGCTTTGGAGATCAGCCTCACCTTCTCCCATCCCAGAAGCGGGGCTTACAGCCAGCCCTTACAGTTTCACTCATGAAGCACCTTGATCTTTGGTGTCCTGGACTTCATCCTGGGTGCTGCAGATAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian-Ping Chen et al.
Molecular medicine (Cambridge, Mass.), 27(1), 21-21 (2021-03-05)
Studies have found that circular RNAs (circRNAs) play key roles in cardiovascular diseases. However, the function of circROBO2 in acute myocardial infarction (AMI) is unclear. This study aimed to investigate the pathogenesis of circROBO2 in AMI. qRT-PCR and Western blot
Hua Wang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 40(5), 1063-1078 (2016-12-14)
Chronic lung infection in cystic fibrosis leads to an inflammatory response that persists because of the chronic presence of bacteria and ultimately leads to a catastrophic failure of lung function. We use a combination of biochemistry, cell and molecular biology
Ganqian Zhu et al.
Journal of immunology (Baltimore, Md. : 1950), 198(3), 1104-1118 (2017-01-01)
The apoptosis of glomerular mesangial cells (GMCs) in the early phase of rat Thy-1 nephritis (Thy-1N), a model of human mesangioproliferative glomerulonephritis (MsPGN), is primarily triggered by sublytic C5b-9. However, the mechanism of GMC apoptosis induced by sublytic C5b-9 remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service