Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU146001

Sigma-Aldrich

MISSION® esiRNA

targeting human P2RX7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCCGTCCCAAATACAGTTTCCGTCGCCTTGACGACAAGACCACCAACGTGTCCTTGTACCCTGGCTACAACTTCAGATACGCCAAGTACTACAAGGAAAACAATGTTGAGAAACGGACTCTGATAAAAGTCTTCGGGATCCGTTTTGACATCCTGGTTTTTGGCACCGGAGGAAAATTTGACATTATCCAGCTGGTTGTGTACATCGGCTCAACCCTCTCCTACTTCGGTCTGGCCGCTGTGTTCATCGACTTCCTCATCGACACTTACTCCAGTAACTGCTGTCGCTCCCATATTTATCCCTGGTGCAAGTGCTGTCAGCCCTGTGTGGTCAACGAATACTACTACAGGAAGAAGTGCGAGTCCATTGTGGAGCCAAAGCCGACATTAAAGTATGTGTCCTTTGTGGATGAATCCCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang Lili et al.
Experimental biology and medicine (Maywood, N.J.), 244(9), 734-742 (2019-05-02)
The mechanism of gastric cancer is highly complex, accompanied by a variety of genetic abnormalities. It is of great significance to elucidate the pathogenesis of gastric cancer, find its markers and therapeutic targets in the fight against this fatal disease.
Hengli Zhao et al.
Scientific reports, 6, 23286-23286 (2016-03-17)
Blockading P2X7 receptor(P2X7R) provides neuroprotection toward various neurological disorders, including stroke, traumatic brain injury, and subarachnoid hemorrhage. However, whether and how P2X7 receptor suppression protects blood-brain barrier(BBB) after intracerebral hemorrhage(ICH) remains unexplored. In present study, intrastriatal autologous-blood injection was used
Shu Guan et al.
Frontiers in psychiatry, 10, 770-770 (2019-11-05)
Diabetic neuropathic pain (DNP) and major depressive disorder (MDD) are common complications of diabetes mellitus and mutually affect each other. As a member of the ATP-gated ion channel family, P2X7 receptor is associated with the transduction of pain signal and
Hui Pan et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(10), 13533-13543 (2016-07-30)
Uveal melanoma (UM) has a high mortality rate for primary intraocular tumors. Approximately half of UM patients present with untreatable and fatal metastases. Long non-coding RNAs (lncRNAs) have emerged as potent regulatory RNAs that play key roles in various cellular
Miso Park et al.
Scientific reports, 9(1), 11587-11587 (2019-08-14)
Tamoxifen (TAM) is the standard anti-hormonal therapy for estrogen receptor-positive breast cancer. However, long-term TAM therapy can make acquisition of TAM resistance and there are still no solutions to treat TAM-resistant breast cancer. In this study, we found that protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service