Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU141161

Sigma-Aldrich

MISSION® esiRNA

targeting human PPM1G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGAGGGAAGCAGTTGATTGTAGCCAACGCAGGAGACTCTCGCTGTGTGGTATCTGAGGCTGGCAAAGCTTTAGACATGTCCTATGATCACAAACCAGAGGATGAAGTAGAACTAGCACGCATCAAGAATGCTGGTGGCAAGGTCACCATGGATGGGCGAGTCAACGGGGGCCTCAACCTCTCCAGAGCCATTGGGGACCACTTCTATAAGAGAAACAAGAACCTGCCACCTGAGGAACAGATGATTTCAGCCCTTCCTGACATCAAGGTGCTGACTCTCACTGACGACCATGAATTCATGGTCATTGCCTGTGATGGCATCTGGAATGTGATGAGCAGCCAGGAAGTTGTAGATTTCATTCAATCAAAGATCAGCCAGCGTGATGAAAATGGGGAGCTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuai Yu et al.
Science advances, 6(47) (2020-11-22)
The adaptor proteins, STING and MAVS, are components of critical pathogen-sensing pathways that induce innate immunity. Phosphorylation of either adaptor results in activation of the type I interferon pathway. How this phosphorylation is regulated and how it is manipulated by
Swapna Aravind Gudipaty et al.
Molecular and cellular biology, 35(22), 3810-3828 (2015-09-02)
Transcription elongation programs are vital for the precise regulation of several biological processes. One key regulator of such programs is the P-TEFb kinase, which phosphorylates RNA polymerase II (Pol II) once released from the inhibitory 7SK small nuclear ribonucleoprotein (snRNP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service