Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU020331

Sigma-Aldrich

MISSION® esiRNA

targeting human IRS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACCCCAATGGCTACATGATGATGTCCCCCAGCGGTGGCTGCTCTCCTGACATTGGAGGTGGCCCCAGCAGCAGCAGCAGCAGCAGCAACGCCGTCCCTTCCGGGACCAGCTATGGAAAGCTGTGGACAAACGGGGTAGGGGGCCACCACTCTCATGTCTTGCCTCACCCCAAACCCCCAGTGGAGAGCAGCGGTGGTAAGCTCTTACCTTGCACAGGTGACTACATGAACATGTCACCAGTGGGGGACTCCAACACCAGCAGCCCCTCCGACTGCTACTACGGCCCTGAGGACCCCCAGCACAAGCCAGTCCTCTCCTACTACTCATTGCCAAGATCCTTTAAGCACACCCAGCGCCCCGGGGAGCCGGAGGAGGGTGCCCGGCATCAGCACCTCCGCCTTTCCACTAGCTCTGGTCGCCTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongxin Luan et al.
International journal of clinical and experimental pathology, 8(9), 10345-10354 (2015-12-01)
MicroRNA (miR-126) was reported to be downregulated and to act as a tumor suppressor in cancers of the lung, cervix, bladder, breast, liver and prostate. However, the precise roles and underling mechanisms of miR-126 in glioma remain largely unknown. This
Y Chen et al.
European review for medical and pharmacological sciences, 23(18), 7989-7999 (2019-10-11)
The important role of microRNA-1271 (miR-1271) has been identified in human diseases and cancers. However, the biological function of miR-1271 remains ambiguous in papillary thyroid carcinoma (PTC). Therefore, the specific role of miR-1271 was investigated in PTC. The expressions of
Peng Wang et al.
Technology in cancer research & treatment, 16(6), 1102-1112 (2018-01-16)
Thyroid cancer is a common endocrine gland malignancy which exhibited rapid increased incidence worldwide in recent decades. This study was aimed to investigate the role of long noncoding RNA H19 in thyroid cancer. Long noncoding RNA H19 was overexpressed or
Qianyi Luo et al.
Investigative ophthalmology & visual science, 60(6), 1928-1936 (2019-05-03)
Diabetes leads to the downregulation of the retinal Kir4.1 channels and Müller cell dysfunction. The insulin receptor substrate-1 (IRS-1) is a critical regulator of insulin signaling in Müller cells. Circadian rhythms play an integral role in normal physiology; however, diabetes
Piero Dalle Pezze et al.
Nature communications, 7, 13254-13254 (2016-11-22)
Amino acids (aa) are not only building blocks for proteins, but also signalling molecules, with the mammalian target of rapamycin complex 1 (mTORC1) acting as a key mediator. However, little is known about whether aa, independently of mTORC1, activate other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service