Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU007261

Sigma-Aldrich

MISSION® esiRNA

targeting human CCP110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGAGCTGCCTTGTACGGTATTCATGACATATTCTTTGTAATGGATGCAGCTGAAAGAATGTCTATTCTACATCATGATCGAGAAGTTCGCAAAGAGAAAATGCTCAGGCAAATGGATAAAATGAAAAGTCCACGAGTGGCTCTTTCAGCTGCAACACAGAAGTCTCTTGATAGGAAGAAATACATGAAAGCTGCTGAAATGGGAATGCCAAATAAGAAATTTCTGGTTAAACAAAATCCTTCTGAAACAAGAGTCCTTCAGCCAAACCAAGGACAGAATGCACCTGTTCATAGGCTACTTAGTAGACAAGGGAGTATATGCAGGAAAAATCCAAAGAAAGCGGCCAAATGTTGCGACAATTTAAGAAGACAACATTCATTAGGATAAAATGGGGGGAAGGATTATTATTCATGTTATTTTCCCTGCCCAAGACTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang Wang et al.
The Journal of cell biology, 218(12), 4030-4041 (2019-10-18)
The primary cilium is a sensory organelle that protrudes from the cell surface. Primary cilia undergo dynamic transitions between assembly and disassembly to exert their function in cell signaling. In this study, we identify the small GTPase Rab7 as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service