E8159
Anti-EDEM1 antibody,Mouse monoclonal
clone EDEM1-17, purified from hybridoma cell culture
Synonym(s):
Anti-EDEM, Anti-ER degradation enhancer, mannosidase alpha-like 1
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
EDEM1-17, monoclonal
form
buffered aqueous solution
mol wt
antigen 75/65 kDa
species reactivity
mouse, human
concentration
~1.0 mg/mL
technique(s)
immunoprecipitation (IP): suitable
western blot: 1-2 μg/mL using whole extracts of HEK-293T cells expressing recombinant human or mouse EDEM1
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... EDEM1(9695)
General description
Application
- immunoblotting
- immunoprecipitation
- immuno-electron microscopy
- immunohistochemistry (IHC)
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service