Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7032

Sigma-Aldrich

EPHB1 (591-end), active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

9330129L11, AW488255, EPHT2, Elk, Elkh, Hek6, Net

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

122-165 nmol/min·mg

mol wt

~62 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Ephb1(270190)

General description

EPHB1 (Ephb1 Eph receptor B1) is a glutamatergic synapse forming tyrosine kinase. Mammals contain five EPHB receptors, namely, EPHB1-EPHB4 and EPHB6. Their ligands are transmembrane in nature and are called ephrinB1, -B2 and -B3.

Application

EPHB1 (591-end), active, GST tagged from mouse has been used for in vitro phosphorylation of Caskin1.

Biochem/physiol Actions

EPHB1 (Ephb1 Eph receptor B1) is a member of the Eph family of receptor tyrosine kinases. Ligand-activated EPHB1 forms a signaling complex with Nck, paxillin, and focal adhesion kinase and induces tyrosine phosphorylation of paxillin in a c-Src-dependent manner to promote cell migration. In addition, activated EPHB1 recruits the adaptor proteins Grb2 and p52Shc and promotes p52Shc and c-Src tyrosine phosphorylation as well as MAPK/extracellular signal-regulated kinase (ERK) activation. Expression of dominant-negative c-Src significantly reduced EPHB1-dependent ERK1/2 activation and chemotaxis. This receptor is essential for the maintenance of hyperalgesia, and is the predominant receptor for chronic pain in the spinal cord.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cécile Vindis et al.
The Journal of biological chemistry, 279(27), 27965-27970 (2004-04-27)
Interactions between Eph receptors and their membrane-bound ligands (ephrins) are of critical importance for key developmental processes such as boundary formation or vascular development. Their downstream signaling pathways are intricate and heterogeneous at several levels, the combined effect being a
Cecile Vindis et al.
The Journal of cell biology, 162(4), 661-671 (2003-08-20)
Eph receptors and their ligands (ephrins) play an important role in axonal guidance, topographic mapping, and angiogenesis. The signaling pathways mediating these activities are starting to emerge and are highly cell- and receptor-type specific. Here we demonstrate that activated EphB1
Vincent Cibert-Goton et al.
PloS one, 8(1), e53673-e53673 (2013-01-24)
EphB receptors tyrosine kinases and ephrinB ligands were first identified as guidance molecules involved in the establishment of topographical mapping and connectivity in the nervous system during development. Later in development and into adulthood their primary role would switch from
Szabolcs Pesti et al.
Cell communication and signaling : CCS, 10(1), 36-36 (2012-11-28)
Scaffold proteins have an important role in the regulation of signal propagation. These proteins do not possess any enzymatic activity but can contribute to the formation of multiprotein complexes. Although scaffold proteins are present in all cell types, the nervous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service