Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK891169109

DURAN® PURE Screw Cap

new

polyethersulfone cap, PTFE coated silicone seal, GL 80 thread

Synonym(s):

891169109, DURE, PURE, cap, closure, lid, packaging, premium, stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121821

material

gray PTFE seal
gray seal
polyethersulfone cap
white cap

Joint

joint threaded cap

feature

autoclavable

packaging

pack of 5 ea

manufacturer/tradename

DWK Life Sciences DURAN® 891169109

cap diam. × H

88 mm × 40 mm

fitting

GL 80 thread

Looking for similar products? Visit Product Comparison Guide

General description

High-quality plastic screw cap with GLS 80® thread size, developed for the needs of the pharmaceutical industry. The DURAN® PURE GLS 80® Screw Cap is made from Polyethersulfone (PESU) with highly inert PTFE coated silicone seal. Designed for use with DURAN® PURE GLS 80® Bottles. The screw cap is compatible with autoclaving and other common sterilization methods. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. The DURAN® PURE GLS 80® Screw Cap is available with the documentation required by regulatory authorities and meets the requirements of the international pharmacopeias.

Application

storage, pharmaceutical packaging, closure, seal

Features and Benefits

  • PSU, Silicon, PTFE
  • High cleanliness due to manufacturing in clean rooms
  • Certificate of Analysis
  • Visual and dimensional inspection
  • 100% traceability of all manufacturing steps
  • Extractables report available
  • A Type III Packaging Material DMF (#28044) for these products has been filed with the FDA. China Center for Drug Evaluation CDE of NMPA registration number B20220000817
  • USP 88 Class VI Pharmaceutical grade materials

Legal Information

DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service