Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK818065408

DURAN® PURE Bottle

new

amber, graduated, neck joint: threaded GL45, dust cover, without cap, bottle capacity 1000 mL

Synonym(s):

818065408, DURAN PURE bottle, DURAN storage bottle, Duran® PURE Bottle, GLS45 bottle, media bottle, pharmaceutical storage bottle, reagent bottle, storage bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121818

material

amber
borosilicate glass 3.3 bottle

Joint

joint: threaded neck GL45

sterility

non-sterile

feature

autoclavable
graduated
without cap
dust cover

packaging

pack of 10 ea

manufacturer/tradename

DWK Life Sciences DURAN® 818065408

bottle capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

High-quality Type I borosilicate glass bottles GL 25 / GL 45, amber coloured from 25 ml to 20 L, developed for the needs of the pharmaceutical industry. DURAN® PURE Bottles are closed with a protective cover to prevent contamination. Very uniform, durable and chemically resistant amber colouring due to the use of innovative technology offers protection for light sensitive compounds. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. DURAN® PURE Bottles are available with the documentation required by regulatory authorities and meet the requirements of the international pharmacopeias.

Application

Media and reagent prep, sterilization, and storage. Culturing vessel. Liquiding handling bottle. Sample storage.

Features and Benefits

  • Protective cover prevents contamination inside the bottle during storage and transport
  • Manufactured from borosilicate glass 3.3
  • Certificate of Analysis (CoA)
  • 100% visual inspection
  • Analysis of hydrolytic resistance according to EP, USP and JP for each batch
  • 100% traceability of all manufacturing steps
  • A Type III Packaging Material DMF (#28042) for these products has been filed with the FDA
  • China Center for Drug Evaluation CDE NMPA registration number B20220000306

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service