Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ND3

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 92090901

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Hybridoma nerve

packaging

tube of 5 μg 92090901-DNA-5UG
pkg of vial of cells 92090901-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat neurone hybrid

Cell Line Description

ND3 hybrid cells were derived after fusion of primary neonatal-rat dorsal-root-ganglion (DRG) neurons with N18Tg2, a mouse neuroblastoma (C1300)-derived azaguanine-resistant line. HAT sensitive hybrids were selected and cloned by limiting dilution. The cells express all isoforms of N-CAM but no embryonic form. Thy 1.1 marker, P31 and PrPc can be detected as well as a weaker expression of F3 and Thy-3.

Application

Study of neuronal differentiation, cell adhesion and expression of sensory neuron specific marker

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 by either aspirating or gently tapping the cells loose; 5% CO2; 37°C. Cells are lightly attached therefore use of trypsin is unnecessary.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service