STEINLIN human cell line
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 88052087, IHW Number: 9087
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human blood
packaging
tube of 5 μg 88052087-DNA-5UG
pkg of vial of cells 88052087-1VL
ethnicity
French
gender
female
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
Looking for similar products? Visit Product Comparison Guide
General description
Consanguineous: No
Cell Line Description
This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.
DNA Profile
A: 101
Cw: 701
B: 801
HLA DRA: 102
HLA DRB1: 301
HLA DRB3: 101
HLA DQAP: 4.1
HLA DQA1: 501
HLA DQB1: 201
HLA DPA1: 1
HLA DPB1: 0301 0401
Cw: 701
B: 801
HLA DRA: 102
HLA DRB1: 301
HLA DRB3: 101
HLA DQAP: 4.1
HLA DQA1: 501
HLA DQB1: 201
HLA DPA1: 1
HLA DPB1: 0301 0401
Serological Profile
HLA A: 1
HLA A ief: 1
HLA C: 7
HLA B: 8
HLA B ief: 8
Bw4/bw6: 6
HLA Dw: 3
HLA DR: 3
HLA Dw2: 24
HLA DR52: 52
HLA DQ: 2
HLA DP: 3 4
HLA A ief: 1
HLA C: 7
HLA B: 8
HLA B ief: 8
Bw4/bw6: 6
HLA Dw: 3
HLA DR: 3
HLA Dw2: 24
HLA DR52: 52
HLA DQ: 2
HLA DP: 3 4
Culture Medium
Subculture Routine
Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service