OX-118
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 13082235-DNA-5UG
pkg of vial of cells 13082235-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
NS1
Reactivity
Human signal regulatory protein gamma (SIRPgamma)
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Description
The hybridoma cell line OX-118 secretes monoclonal IgG1 antibodies reactive with human signal regulatory protein gamma but it cross reacts with SIRPalpha.
Immunogen
Recombinant human signal regulatory protein gamma (SIRPgamma)
Application
Detection of SIRPgamma (but note cross reaction with SIRPalpha).
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS)
Subculture Routine
Seed at 5 x 104 cells/ml (5 x 105 cells/ml at resuscitation). Subculture when cells reach 5 x 105 - 1 x 106 cells/ml (usually after 3-4 days), 5% CO2, 37°C. 20% FBS can be used at resuscitation.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service