Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KARPAS 384

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Synonym(s):

KARPAS, 384, KARPAS-384, KARPAS384

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

packaging

tube of 5 μg 06120602-DNA-5UG
pkg of vial of cells 06120602-1VL

growth mode

Suspension

karyotype

Complex, refer to Dyer et al., 1993

morphology

Large clumps growing slowly in suspension

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human T-cell non-Hodgkin lymphoma

Cell Line Description

Established from peripheral blood of a 48 year-old male with widespread subcutaneous nodules. Biopsy showed high grade Non-Hodgkin lymphoma with numerous mitoses.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Doubling time approximately 60 hours. Cells grow slowly in suspension in large clumps. Maintain cultures between 3-9 x 105 cells/ml; 5% CO2; 37 °C. Thaw ampoule and perform a cell count, spin out the DMSO and resuspend in an appropriate volume of media to give a cell concentration of 4-5 x 105 cells/ml. Viability may be poor on resuscitation and may decrease further within the first 48 hours, the culture may appear non-viable at this time. Clusters of cells will eventually appear but this could take 1-2 weeks, during this time large media changes are not recommended. It may take 2-3 weeks for vigorous growth to be established. At subculture perform a cell count and maintain cultures between 3-9 x 105 cells/ml, 5% CO2, 37 °C. For cryopreservation of KARPAS 384, freeze at approximately 2 x 107 cells/ml in 90% FBS + 10% DMSO.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service