OX-101
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 03062503-DNA-5UG
pkg of vial of cells 03062503-1VL
ImmunoDon
BALB/c mouse
Myeloma
NS1
Reactivity
Anti-CD47, rat
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS1 x mouse BALB/c
Cell Line Description
This antibody recognises the widely distributed CD47 (integrin associated protein), the ligand of SIRPα.
Immunogen
Biotinylated rat (AO) thymocytes
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Seed at 5 x 10,000 cells/ml. Subculture when cells reach 500,000 - 1,000,000 cells/ml (usually after 3-4 days) 5% CO2 37° C. Start in 20% FBS when recovering from frozen.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service