Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9369

Sigma-Aldrich

CCT007093

≥98% (HPLC), powder, yellow

Synonym(s):

(2E,5E)-2,5-Bis(2-thienylmethylene)-cyclopentanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12OS2
CAS Number:
Molecular Weight:
272.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder, yellow

solubility

DMSO: >3 mg/mL

storage temp.

2-8°C

SMILES string

O=C1\C(CC\C1=C/c2cccs2)=C\c3cccs3

InChI

1S/C15H12OS2/c16-15-11(9-13-3-1-7-17-13)5-6-12(15)10-14-4-2-8-18-14/h1-4,7-10H,5-6H2/b11-9+,12-10+

InChI key

KPFZCKDPBMGECB-WGDLNXRISA-N

Application

CCT007093 has been used as a wild-type p53-induced phosphatase (Wip1) inhibitor to study its effect on skin keratinocytes, under ultra-violet (UV) stress conditions. It has also been used as a Wip1 inhibitor in time course inhibition assay to evaluate the effect of Wip1 inhibition on expression of AKT serine/threonine kinase 1(Akt1) and glycogen synthase kinase-3β (GSK-3β)and the phosphorylation of GSK-3β at serine 9 (Ser9) and Akt at Ser473.

Biochem/physiol Actions

CCT007093 inhibits the growth of cancer cell lines (MCF-7, KPL-1, and MCF-3B) that overexpress protein phosphatase, Mg2+/Mn2+ dependent 1D (PPM1D). It induces cancer cell death via the activation of p38 kinase activity.
CCT007093 is an effective PPM1D inhibitor that selectively reduces viability of human tumour cell lines.
CCT007093 is an effective protein phosphatase, Mg2+/Mn2+ dependent, 1D (PPM1D) inhibitor that selectively reduces viability of human tumour cell lines.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Serine/Threonine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Yin et al.
Oncotarget, 7(20), 29359-29370 (2016-04-29)
Inactivation of p53 greatly contributes to serous ovarian cancer, while the role of the wild-type p53 induced phosphatase 1 (Wip1) is quite unclear. In this study, by silencing or overexpression of Wip1, we found that Wip1 suppressed ovarian cancer cell
Joshua A Bauer et al.
Breast cancer research : BCR, 12(3), R41-R41 (2010-06-26)
Paclitaxel is a widely used drug in the treatment of patients with locally advanced and metastatic breast cancer. However, only a small portion of patients have a complete response to paclitaxel-based chemotherapy, and many patients are resistant. Strategies that increase
S Rayter et al.
Oncogene, 27(8), 1036-1044 (2007-08-19)
The PPM1D gene is aberrantly amplified in a range of common cancers and encodes a protein phosphatase that is a potential therapeutic target. However, the issue of whether inhibition of PPM1D in human tumour cells that overexpress this protein compromises
J-Y Song et al.
Cell death & disease, 4, e744-e744 (2013-08-03)
Wild-type p53-induced phosphatase 1 (Wip1) is a p53-inducible serine/threonine phosphatase that switches off DNA damage checkpoint responses by the dephosphorylation of certain proteins (i.e. p38 mitogen-activated protein kinase, p53, checkpoint kinase 1, checkpoint kinase 2, and uracil DNA glycosylase) involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service