Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C7112

Sigma-Aldrich

Anti-Cortactin (KE-20) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-EMS1, Anti-p80/85

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse, chicken

technique(s)

indirect immunofluorescence: 1:500 using mouse fibroblasts NIH3T3 cell line
western blot: 1:1,000 using cytosol extract of rat brain, whole extract of chicken embryo fibroblasts, or whole cell extract of the human epidermoid carcinoma A431 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTTN(2017)
mouse ... Cttn(13043)
rat ... Cttn(60465)

General description

Cortactin is an F-actin binding protein. It belongs to a highly conserved family of perimembrane signaling protein. Cortactin contains a unique multi-domain structure consisting of 6.5 tandem repeats of a 37-amino acid sequence required for direct binding to F-actin, followed by an α-helix region, a proline-rich region, and three tyrosine residues targeted by Src related kinases. The C-terminal region contains a Src-homology 3 (SH3) domain.
Cortactin is encoded by the CTTN gene on chromosome 11 in humans. It is expressed in most adherent cells and is a prominent substrate of protein-tyrosine kinase Src. The protein localizes within lamellipodia and at the cell surface. Cortactin is also called as EMS1.

Immunogen

synthetic peptide corresponding to the mid-region of human cortactin (amino acids 361-380) conjugated to KLH.

Application

Anti-Cortactin (KE-20) antibody produced in rabbit is suitable for indirect immunofluorescence at a dilution of 1:500 using mouse fibroblasts NIH3T3 cell line and western blotting at a dilution of 1:1,000 using cytosol extract of rat brain, whole extract of chicken embryo fibroblasts, or whole cell extract of the human epidermoid carcinoma A431 cell line.

Biochem/physiol Actions

Cortactin is a cytoskeleton protein that facilitates the assembly of cortical actin. It is activated by phosphorylation of tyrosine kinases or serine/threonine kinases. It plays an important role in promoting lamellipodia formation, invadopodia formation, cell migration, and endocytosis. Cortactin regulates the interactions between components of adherens-type junctions and helps in organizing the cytoskeleton, cell adhesion structures of epithelia and carcinoma cells. During apoptosis, the encoded protein is degraded in a caspase-dependent manner. It acts as a regulator in invasion and migration of tumor cells. Overexpression of cortactin causes osteosarcoma carcinogenesis and along with FAK increases the risk of oral premalignant lesions.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of Arp2/3 complex-mediated actin polymerization by cortactin
Uruno T, et al.
Nature Cell Biology, 3(3), 259-259 (2001)
Arlo Adams et al.
Biology of reproduction, 103(6), 1314-1323 (2020-09-10)
The release of late spermatids from the seminiferous epithelium requires the internalization of intercellular junctions by Sertoli cell specific structures called "tubulobulbar complexes" (TBCs). These large, endocytic devices likely evolved from classic clathrin-mediated-endocytosis (CME) machinery, but have several important morphological
David Hattan et al.
The Journal of biological chemistry, 277(41), 38596-38606 (2002-08-02)
Tyrosine phosphorylation evokes functional changes in a variety of ion channels. Modulation of the actin cytoskeleton also affects the function of some channels. Little is known about how these avenues of ion channel regulation may interact. We report that the
Aviv Bergman et al.
Cell adhesion & migration, 8(3), 273-279 (2014-04-10)
Invadopodia are dynamic protrusions in motile tumor cells whose function is to degrade extracellular matrix so that cells can enter into new environments. Invadopodia are specifically identified by microscopy as proteolytic invasive protrusions containing TKS5 and cortactin. The increasing complexity
Cecilia Folio et al.
Cancer biomarkers : section A of Disease markers, 10(1), 35-41 (2012-02-03)
The cortactin (CTTN) gene has been found, by transcriptomic profiling, to be overexpressed in pediatric osteosarcoma. The location of CTTN at 11q13 and the role of cortactin in cytoskeleton restructuring make CTTN of interest as a potential biomarker for osteosarcoma.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service