Key Documents
BR701432
BRAND® Life Science heat sealing film
new
long term storage, aluminum film (adhesive backed), operating temperature (-200)—110 °C, pack of 100 ea
Synonym(s):
701432, BRAND film, aluminum film, cover, heat sealing, plate film, seal, sealing film, storage film
About This Item
UNSPSC Code:
41122410
material
aluminum film (adhesive backed)
sterility
non-sterile
feature
DNA free
RNase and DNase free
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701432
parameter
(-200)—110 °C operating temperature
General description
Heat sealing films provide protection against evaporation and contamination. Aluminum foils are used for PP and PS plates. They are resistant to solvents and are used both at low temperatures and for storage at room temperature. Depending on the application, they are peelable or pierceable.
Features and Benefits
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant
  • Peelable and pierceable
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)