Key Documents
BR701431
BRAND® Life Science heat sealing film
new
qPCR, suitable for PCR, storage, PE/PP/adhesive, operating temperature (-80)—110 °C, pack of 100 ea, transparent
Synonym(s):
701431, BRAND film, PE/PP film, cover, heating sealing, plate film, seal, sealing film, storage film
About This Item
UNSPSC Code:
41122410
material
polyethylene film
polypropylene film (adhesive backed)
sterility
non-sterile
feature
DNA free
RNase and DNase free
qPCR
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701431
parameter
(-80)—110 °C operating temperature
suitability
suitable for PCR
General description
Heat sealing films provide protection against evaporation and contamination. Aluminum foils are used for PP and PS plates. They are resistant to solvents and are used both at low temperatures and for storage at room temperature. Depending on the application, they are peelable or pierceable.
Features and Benefits
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant
  • Peelable and pierceable
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)