Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPKF384RS

Pipette filter tips for Packard® plate track

capacity 30 μL, clear, racked, pkg of 5x3840tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
30 μL
sterility:
sterile
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc PKF-384-R-S
Corning

sterility

sterile

manufacturer/tradename

Axygen Inc PKF-384-R-S
Corning

packaging

pkg of 5x3840tips/cs

capacity

30 μL

volume

30 μL

color

clear

Other Notes

Browse all pipette tips

Legal Information

Packard is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service