Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV48521

Sigma-Aldrich

Anti-FNTA antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FPTA, Anti-Farnesyltransferase, CAAX box, α, Anti-MGC99680, Anti-PGGT1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FNTA(2339)

General description

Farnesyltransferase, CAAX box, α (FNTA) forms a part of the heterodimeric CAAX farnesyltransferase complex that attaches a farnesyl moiety to a cysteine residue in proteins. The protein substrates usually comprise of nuclear lamins, retinal proteins and ras proteins.
Rabbit Anti-FNTA antibody recognizes canine, rat, human, and bovine FNTA.

Immunogen

Synthetic peptide directed towards the C terminal region of human FNTA

Application

Rabbit Anti-FNTA antibody is suitable for western blot applications at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

Prenyltransferases attach either a farnesyl group or a geranylgeranyl group in thioether linkage to the cysteine residue of protein′s with a C-terminal CAAX box. CAAX geranylgeranyltransferase and CAAX farnesyltransferase are heterodimers that share the same alpha subunit but have different beta subunits. FNTA is the alpha subunit of these transferases.Prenyltransferases attach either a farnesyl group or a geranylgeranyl group in thioether linkage to the cysteine residue of protein′s with a C-terminal CAAX box. CAAX geranylgeranyltransferase and CAAX farnesyltransferase are heterodimers that share the same alpha subunit but have different beta subunits. This gene encodes the alpha subunit of these transferases. Alternative splicing results in multiple transcript variants encoding different isoforms.

Sequence

Synthetic peptide located within the following region: DNKEDILNKALELCEILAKEKDTIRKEYWRYIGRSLQSKHSTENDSPTNV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Andres et al.
Genomics, 18(1), 105-112 (1993-10-01)
The CAAX farnesyltransferase is a heterodimeric enzyme that attaches a farnesyl group to a single cysteine in several cellular proteins. Substrates include the p21ras proteins, nuclear lamins, and several retinal proteins, all of which end with a "CAAXbox," where C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service