Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV36976

Sigma-Aldrich

Anti-SIN3B antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

105 kDa

species reactivity

mouse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... SIN3B(20467)

Immunogen

Synthetic peptide directed towards the N terminal of human SIN3B

Biochem/physiol Actions

Sin3B is one of the Sin3 co-repressors act as a protein scaffold to recruit transcription factors via its four highly homologous paired amphipathic helix (PAH) domains.

Sequence

Synthetic peptide located within the following region: NIQSPLSSQDNSHSHGDCGEDFKQMSYKEDRGQVPLESDSVEFNNAISYV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Zheng et al.
Endocrinology, 155(11), 4507-4520 (2014-07-31)
Endocrine regulation of uterine biology is critical for embryo receptivity and human reproduction. Uterine endometrium depends on extrinsic sex steroid input and hence likely has mechanisms that enable adaptation to hormonal variation. Emerging evidence suggests that sex steroid bioavailability in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service