Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV34684

Sigma-Aldrich

Anti-MEIS2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Meis homeobox 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

bovine, rabbit, human, guinea pig, horse, dog, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEIS2(4212)

General description

MEIS2 is a homeobox protein that is involved in the development of lens and retina. It is also known to compete with Tle4 for Otx2 binding and modulates tectal fate.
Rabbit Anti-MEIS2 antibody recognizes chicken, human, mouse, rat, zebrafish, canine, and bovine MEIS2.

Immunogen

Synthetic peptide directed towards the N terminal region of human MEIS2

Application

Rabbit Anti-MEIS2 antibody is suitable for western blot (1.25 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

MEIS2 encodes a homeobox protein belonging to the TALE (′three amino acid loop extension′) family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs.

Sequence

Synthetic peptide located within the following region: HYGAHAPHPNVMPASMGSAVNDALKRDKDAIYGHPLFPLLALVFEKCELA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zsuzsa Agoston et al.
Development (Cambridge, England), 136(19), 3311-3322 (2009-09-09)
The transcription factor Otx2 is expressed throughout the anterior neuroectoderm and is required for the formation of all forebrain- and midbrain-derived structures. The molecular determinants that cooperate with Otx2 to subdivide its expression domain into distinct functional units are, however
Ivan Conte et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(35), 15491-15496 (2010-08-18)
MicroRNAs (miRNAs) are small noncoding RNAs that have important roles in the regulation of gene expression. The roles of individual miRNAs in controlling vertebrate eye development remain, however, largely unexplored. Here, we show that a single miRNA, miR-204, regulates multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service