Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV33823

Sigma-Aldrich

Anti-FABP3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FABP3(2170)

Immunogen

Synthetic peptide directed towards the N terminal region of human FABP3

Biochem/physiol Actions

Fatty acid-binding protein 3 (FABP3) gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is a candidate tumor suppressor gene for human breast cancer.

Sequence

Synthetic peptide located within the following region: NGDILTLKTHSTFKNTEISFKLGVEFDETTADDRKVKSIVTLDGGKLVHL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Zhao et al.
Lipids, 44(11), 1029-1037 (2009-10-23)
Intramuscular fat (IMF) content affects meat quality and varies in different pig breeds. However, the underlying mechanisms of different IMF depositions in different genetic backgrounds of pigs have not been fully elucidated as yet. Lipid metabolism theoretically contributes to the
Ariful Islam et al.
The Journal of nutrition, 144(10), 1509-1516 (2014-08-15)
Low placental fatty acid (FA) transport during the embryonic period has been suggested to result in fetal developmental disorders and various adult metabolic diseases, but the molecular mechanism by which FAs are transported through the placental unit remains largely unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service