Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV02014

Sigma-Aldrich

Anti-SEPT4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Septin-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Sept4 is a part of the Lewy body complexes that supress α-synuclien toxicity. Furthermore, Sept4 has been implicated in Parkinson′s disease, stem cell death and tumor supression.
Rabbit Anti-SEPT4 antibody binds to canine, and human Sept4.

Immunogen

Synthetic peptide directed towards the N terminal region of human SEPT4 (septin-4)

Application

Rabbit Anti-SEPT4 antibody can be used for IHC (4-8μg/ml, using paraffin-embedded tissues) and western blot (0.5μg/ml) applications.

Sequence

Synthetic peptide located within the following region: RSLGWQGNSVPEDRTEAGIKRFLEDTTDDGELSKFVKDFSGNASCHPPEA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masafumi Ihara et al.
Neuron, 53(4), 519-533 (2007-02-14)
In Parkinson disease (PD), alpha-synuclein aggregates called Lewy bodies often involve and sequester Septin4 (Sept4), a polymerizing scaffold protein. However, the pathophysiological significance of this phenomenon is unclear. Here, we show the physiological association of Sept4 with alpha-synuclein, the dopamine
María García-Fernández et al.
Genes & development, 24(20), 2282-2293 (2010-10-19)
Inhibitor of Apoptosis Proteins (IAPs) are frequently overexpressed in tumors and have become promising targets for developing anti-cancer drugs. IAPs can be inhibited by natural antagonists, but a physiological requirement of mammalian IAP antagonists remains to be established. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service