Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

280R-2

Sigma-Aldrich

Mammaglobin (EP249) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP249, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (280R-24)
vial of 0.5 mL concentrate (280R-25)
bottle of 1.0 mL predilute (280R-27)
vial of 1.0 mL concentrate (280R-26)
bottle of 7.0 mL predilute (280R-28)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

breast carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... SCGB2A2(4250)

General description

Mammaglobin (10 kD) is a breast-associated glycoprotein distantly related to secretoglobin family that includes human uteroglobin and lipophilin. Mammaglobin mRNA is present in high levels in human breast cancer cell lines, which has been shown to be a sensitive marker of breast cancer.1,2,3 When combined with other breast-restricted markers such as GCDFP-15, an overall sensitivity for breast carcinoma of 84% has been achieved.4 Anti-mammaglobin can also be used to determine breast origin in the setting of metastatic carcinoma.5,6

Quality


IVD

IVD

IVD

RUO

Linkage

Mammaglobin Positive Control Slides, Product No. 280S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T Fremeau et al.
Brain research. Molecular brain research, 6(2-3), 197-201 (1989-11-01)
We describe here the in situ hybridization procedure which we have used to detect the pro-opiomelanocortin (POMC) gene primary transcript in nuclei of individual neurons in the periarcuate region of the hypothalamus. An exon-intron RNA probe was used to detect
Jae-Ho Han et al.
Archives of pathology & laboratory medicine, 127(10), 1330-1334 (2003-10-03)
Organ specificity is a desirable property of a tumor marker, especially in metastatic adenocarcinomas of unknown primary origin. Mammaglobin, a mammary-specific member of the uteroglobin family, is known to be overexpressed in human breast cancer. We investigated mammaglobin A expression
T P Fleming et al.
Annals of the New York Academy of Sciences, 923, 78-89 (2001-02-24)
The mammaglobin gene encodes a 10-kDa glycoprotein that is distantly related to a family of proteins that includes rat estramustine binding protein (EMBP)/prostatein and human Clara cell 10-kDa protein (CC10)/uteroglobin. Among normal adult tissues, mammaglobin mRNA expression has been detected
Eiichi Sasaki et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 20(2), 208-214 (2006-12-29)
Previously, we used the reverse transcription-polymerase chain reaction (RT-PCR) to show that mammaglobin (MGB1) can serve as a differential marker of breast cancer metastasis from primary lung cancer. However, mRNA-based methods are not appropriate for use in clinical practices. In
M A Watson et al.
Cancer research, 59(13), 3028-3031 (1999-07-09)
The mammaglobin gene encodes a novel, breast cancer-associated glycoprotein. In this study, we have evaluated the frequency with which mammaglobin expression can be detected in primary and metastatic breast tumors and in breast tumor cells present in the peripheral circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service