Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33538

Sigma-Aldrich

1-Propanol

puriss. p.a., reag. Ph. Eur., ≥99.5% (GC)

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
technique(s):
GC/GC: suitable
bp:
97 °C (lit.)
vapor pressure:
10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Agency

USP/NF
reag. Ph. Eur.

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

autoignition temp.

700 °F

expl. lim.

13.7 %

technique(s)

GC/GC: suitable

impurities

≤0.001% free acid (as C2H5COOH)
≤0.001% non-volatile matter
≤0.01% acetone (GC)
≤0.01% propionaldehyde
≤0.05% 2-propanol
≤0.05% ethanol (GC)
≤0.05% methanol (GC)
≤0.05% water (Karl Fischer)

refractive index

n20/D 1.384 (lit.)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Propanol is a linear alcohol. It exhibits Debye-type relaxation process which resembles α-relaxation and Johari-Goldstein type β-relaxation features of supercooled liquids (nonhydrogen-bonding type). It has been tested as a solvent for direct methanol/oxygen fuel cells.

Application

1-Propanol has been employed as a solvent for the synthesis of mesoporous alumina.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous alumina (I): comparison of synthesis schemes using anionic, cationic, and non-ionic surfactants.
Ray JC, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 100(1), 183-190 (2007)
Dynamics of glass-forming liquids. III. Comparing the dielectric α-and β-relaxation of 1-propanol and o-terphenyl.
Hansen C, et al.
J. Chem. Phys., 107, 1086-1093 (1997)
Evaluation of Ethanol, 1-Propanol, and 2-Propanol in a Direct Oxidation Polymer-Electrolyte Fuel Cell A Real-Time Mass Spectrometry Study.
Wang J, et al.
Journal of the Electrochemical Society, 142(12), Wang J-Wang J (1995)
David Fernandez Rivas et al.
Ultrasonics sonochemistry, 19(6), 1252-1259 (2012-05-23)
Micromachined pits on a substrate can be used to nucleate and stabilize microbubbles in a liquid exposed to an ultrasonic field. Under suitable conditions, the collapse of these bubbles can result in light emission (sonoluminescence, SL). Hydroxyl radicals (OH()) generated
Takeshi Morita et al.
The journal of physical chemistry. B, 116(24), 7328-7333 (2012-06-01)
We characterized the effects of ethanol (ET) and dimethyl sulfoxide (DMSO) on H(2)O within a limited H(2)O-rich region by the 1-propanol (1P)-probing methodology developed by us earlier. The results are displayed on a two-dimensional map with twin coordinates: one pertaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service