Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000709

Oxitropium bromide

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26BrNO4
CAS Number:
Molecular Weight:
412.32
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxitropium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].[N+]1(C2C3OC3C1CC(C2)OC(=O)C(CO)c4ccccc4)(CC)C

InChI

1S/C19H26NO4.BrH/c1-3-20(2)15-9-13(10-16(20)18-17(15)24-18)23-19(22)14(11-21)12-7-5-4-6-8-12;/h4-8,13-18,21H,3,9-11H2,1-2H3;1H/q+1;/p-1

InChI key

LCELQERNWLBPSY-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxitropium bromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takanobu Shioya et al.
Arzneimittel-Forschung, 58(1), 24-28 (2008-03-29)
The present study was undertaken to evaluate the long-term effect of procaterol hydrochloride (CAS 62929-91-3, Meptin), a third generation beta2-receptor agonist on lung function, exercise capacity, health-related quality of life (HRQOL) and activities of daily living (ALDs) in patients with
Nicholas J Gross
European journal of pharmacology, 533(1-3), 36-39 (2006-02-21)
Anticholinergic agents have important uses as bronchodilators for the treatment of obstructive airway diseases, both asthma and, more particularly, chronic obstructive pulmonary disease (COPD). Those in approved clinical use are synthetic quaternary ammonium congeners of atropine, and include ipratropium bromide
Yutaka Nakano et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 90(3), 331-337 (2003-04-03)
International guidelines recommend multiple doses of inhaled beta2-agonists and anticholinergics plus early administration of systemic corticosteroids for acute, severe asthma. This study examined the efficacy of this protocol in adults and analyzed those factors associated with unresponsiveness to the protocol
D C McCrory et al.
The Cochrane database of systematic reviews, (4)(4), CD003900-CD003900 (2003-01-10)
Inhaled bronchodilators form the mainstay of treatment for acute exacerbations of COPD. Two types of agent are used routinely, either singly or in combination: anticholinergic agents and beta2-sympathomimetic agonists. To assess the effect of anti-cholinergic agents on lung function and
Cristoforo Incorvaia et al.
Respiratory medicine, 101(3), 476-480 (2006-08-22)
Inhaled bronchodilators are first line drugs in the treatment of chronic obstructive pulmonary disease (COPD). Tiotropium bromide is a recently introduced long-acting anticholinergic agent able to reduce dyspnoea and COPD exacerbations and to improve pulmonary function and quality of life.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service