Y0000431
Trospium impurity B
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(1R,3r,5S)-8-Azabicyclo[3.2.1]oct-3-yl hydroxydiphenylacetate
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
trospium
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
N1C2CCC1CC(C2)OC(=O)C(O)(c4ccccc4)c3ccccc3
InChI
1S/C21H23NO3/c23-20(25-19-13-17-11-12-18(14-19)22-17)21(24,15-7-3-1-4-8-15)16-9-5-2-6-10-16/h1-10,17-19,22,24H,11-14H2
InChI key
HIIVXBCWDPCZJA-UHFFFAOYSA-N
General description
Application
Packaging
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service