Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UPTQBF1

Sigma-Aldrich

Ultra pure 10x Taq Buffer with MgCl2

DNA-free, Nuclease-free 10x PCR reaction buffer with MgCl2, designed for further manufacture

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

form

liquid

feature

DNA free
RNase and DNase free (also Nickase free)

packaging

pkg of 1.5 mL

technique(s)

RT-PCR: suitable
cDNA synthesis: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Ultra pure 10x Taq Buffer with MgCl2 is manufactured under stringent conditions to ensure that it is free of detectable levels of DNA, DNase, RNase, and Nickase to reduce the risk of false positive and false negative results in molecular diagnostic assays. Ultra pure 10x Taq Buffer with MgCl2 should be used with other ultra pure DNA-free reagents, such as Ultra pure JumpStart Taq DNA Polymerase and Ultra pure DNA-free Water, to minimize false positive and negative results and improved DNA amplification.
Ultra pure 10x Taq Buffer with MgCl2 was designed for use in highly-regulated industries in applications that require a high level of performance, lot-to-lot consistency, and high sensitivity without false results from reagent-borne contamination while meeting industry-driven critical regulations. To meet this requirements we have increased the quality level of our products. The M-Clarity Program defines product quality. It provides transparency so that you can choose, with confidence, suitable products that meet your needs with respect to quality and regulatory compliance, transparency, change notification, and documentation.

Application

Ultra pure 10x Taq Buffer with MgCl2 can be used for:
  • Quantitative PCR
  • Amplification of complex genomic DNA and cDNA templates
  • Amplification of low-copy number targets
  • Microbiome analysis
  • For further manufacture

Features and Benefits

  • DNA-free
  • DNase-free, RNase-free, and nickase-free
  • Ideal for high throughput quantitative PCR applications
  • Compatible with Ultra pure JumpStart Taq DNA Polymerase
  • Enhanced change control and quality agreement

Packaging

Ultra pure 10x Taq Buffer with MgCl2 is provided in larger SKU sizes for ease of use in high throughput PCR applications and for further manufacture. Ultra pure 10x Taq Buffer with MgCl2 should be used with other ultra pure DNA-free reagents, such as Ultra pure JumpStart Taq DNA Polymerase and Ultra pure DNA-free Water to minimize false positive and negative results.

Other Notes

Quality testing ensures the absence of detectable levels of contaminating DNA (<=1 copy of prokaryotic gDNA, <= 0.1 copy of eukaryotic gDNA, and <=10 copies of plasmid DNA per 100 units of enzyme) as detected by qPCR using universal primers against 16S capable of detecting > 500,000 prokayotes, 18S capable of detecting > 110,000 eukaryotes, and Ori capable of detecting > 25,000 plasmids, respectively.

Legal Information

JumpStart is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service