Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0307000

Penbutolol sulfate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60N2O8S
CAS Number:
Molecular Weight:
680.94
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

penbutolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N(C[C@H](O)COc3c(cccc3)C4CCCC4)C(C)(C)C.N(C[C@H](O)COc1c(cccc1)C2CCCC2)C(C)(C)C

InChI

1S/2C18H29NO2.H2O4S/c2*1-18(2,3)19-12-15(20)13-21-17-11-7-6-10-16(17)14-8-4-5-9-14;1-5(2,3)4/h2*6-7,10-11,14-15,19-20H,4-5,8-9,12-13H2,1-3H3;(H2,1,2,3,4)/t2*15-;/m00./s1

InChI key

FEDSNBHHWZEYTP-ZFQYHYQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Penbutolol sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Funato et al.
Journal of chromatography. B, Biomedical sciences and applications, 757(2), 229-235 (2001-06-22)
This paper describes a new method of high-performance liquid chromatography with chemiluminescence detection for the analysis of penbutolol (PB) and its main metabolite, 4-hydroxy penbutolol (4-OH PB) in rat plasma. 4-Dimethylaminosulfonyl-7-(N-chloroformylmethyl-N-methyl) amino-2,1,3-benzoxadiazole (DBD-COCl) was used as a fluorogenic labeling reagent.
S Hjorth et al.
The Journal of pharmacology and experimental therapeutics, 265(2), 707-712 (1993-05-01)
Recently, we found that the beta 1/beta 2 adrenoceptor blocking agent (-)penbutolol prevents behavioral and biochemical actions of the specific serotonin (5-HT)1A agonist (+/-)-8-hydroxy-2-(di-n-propylamino)tetralin. The putative 5-HT1 receptor antagonist profile of (-)penbutolol was further explored in the present study, using
S Hjorth et al.
Brain research, 835(2), 224-228 (1999-07-23)
Serotonin (5-hydroxytryptamine, 5-HT) autoreceptors may desensitize during prolonged administration of antidepressant drugs. If autoreceptors desensitize, their inhibitory influence on extracellular 5-HT should be attenuated. To test this hypothesis, the selective serotonin reuptake inhibitor (SSRI) citalopram (10 mg kg(-1), s.c., b.i.d.)
E P Korovina
Klinicheskaia meditsina, 76(12), 30-35 (1999-03-06)
Effects of beta-blockers (propranolol, penbutolol) and calcium antagonists (nifedipine, verapamil, diltiazem) were studied in 73 patients with hypertrophic cardiomyopathy (HC). Clinical data, ECG and echo-CG findings were assessed. It was found that beta-adrenoblockers and calcium antagonists improve quality of life
L Shen et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 30(9), 702-705 (1995-01-01)
A method for the detection and identification of penbutolol and its metabolites in human body fluid by using gas chromatography-mass spectrometry was established. The urine was extracted with diethyl ether-isopropanol and the plasma was extracted with Sep-Pak column. After TMS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service