CRM8021
Instrument Calibration Standard 3
for ICP-MS, certified reference material, 1000 μg/mL each component in 5% HNO3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
bottle of 125 mL
concentration
1000 μg/mL each component in 5% HNO3
format
multi-component solution
storage temp.
room temp
General description
For preparation every two weeks or as needed. Dilute to the concentration appropriate for the instrument with equal parts of Nitric Acid Blank and Water Blank.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Analyte
Description
Ca 1000 μg/mL
Fe 1000 μg/mL
K 1000 μg/mL
Mg 1000 μg/mL
Na 1000 μg/mL
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service