BCR185R
Bovine liver (trace elements)
BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
agriculture
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
storage temp.
2-8°C
Analysis Note
For more information please see:
BCR185R
BCR185R
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Trace Elements: Zinc Zn), Selenium (Se), Lead (Pb), Manganese (Mn), Copper (Cu), Cadmium (Cd), Arsenic (As)
Matrix Group: Meat
Trace Elements: Zinc Zn), Selenium (Se), Lead (Pb), Manganese (Mn), Copper (Cu), Cadmium (Cd), Arsenic (As)
Matrix Group: Meat
Sales restrictions may apply
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marcin Brzeziński et al.
Ecotoxicology (London, England), 23(5), 767-778 (2014-02-28)
Variations in the contaminant burden in feral and ranch mink, resulting from differences in their diet, may permit the identification of farm escapees. However, this is only possible in the case of contaminants that accumulate to significantly different levels in
Claudia E Lazarte et al.
Food & nutrition research, 59, 27796-27796 (2015-12-03)
Zinc deficiency is a significant problem, in developing countries and in vegetarians, which can be caused by plant-based diets. Thus, dietary strategies, such as fermentation, to improve zinc bioavailability of diets should be investigated. To improve zinc bioavailability in a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service