Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936367

Sigma-Aldrich

Ethyl acetate

new
greener alternative

BioRenewable, ≥99.5%

Synonym(s):

EtOAc, Ethyl ester, Ethyl ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12352200
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

colorless

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to providing environmentally friendly products, and Bio Ethyl Acetate is a prime example of our dedication to this effort. It is a bio-based solvent derived from renewable sources, which makes it a sustainable drop-in replacement alternative to petroleum-based ethyl acetate. Bio Ethyl Acetate aligns with the principles of "Use of Renewable Feedstocks and is an excellent option for companies and researchers seeking to reduce their environmental footprint while still delivering high-quality products.

Application

- Widely used as a solvent for the extraction of various compounds from natural sources

-Analytical chemistry
oThin-layer chromatography (TLC)
oFlash chromatography

-Chemical Synthesis
oEsterifications
oAcylations
oCondensations

Features and Benefits

  • Made from Renewable Resource
  • Replacement for Petroleum-based ethyl acetate
  • Renewable Carbon Content - ≥99.9%
  • ASTM D6866 - Standard Test Methods for Determining the Biobased Content

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service